missing translation for 'onlineSavingsMsg'
Learn More

trans-4-Nitrocinnamic acid, 98+%

Catalog No. AAA1299014
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1299014 25 g
AAA1299022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1299014 Supplier Thermo Scientific Chemicals Supplier No. A1299014

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 882-06-4 | C9H7NO4 | 193.158 g/mol

trans-4-Nitrocinnamic acid, is used as a raw material for saccharine. It is also used as a useful chemical for biological applications. Some other applications include it is used as an organic intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-4-Nitrocinnamic acid, is used as a raw material for saccharine. It is also used as a useful chemical for biological applications. Some other applications include it is used as an organic intermediate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong bases.

Chemical Identifiers

CAS 882-06-4
Molecular Formula C9H7NO4
Molecular Weight (g/mol) 193.158
MDL Number MFCD00007381
InChI Key XMMRNCHTDONGRJ-ZZXKWVIFSA-N
Synonym 4-nitrocinnamic acid, 3-4-nitrophenyl acrylic acid, p-nitrocinnamic acid, cinnamic acid, p-nitro, 2-propenoic acid, 3-4-nitrophenyl, e-3-4-nitrophenyl acrylic acid, 3-4-nitrophenyl propenoic acid, trans-4-nitrocinnamic acid, ccris 1665, 2e-3-4-nitrophenyl prop-2-enoic acid
PubChem CID 737157
SMILES C1=CC(=CC=C1C=CC(=O)O)[N+](=O)[O-]

Specifications

Melting Point ∼290°C (decomposition)
Odor Odorless
Quantity 25 g
Beilstein 1369659
Solubility Information Insoluble in water.
Formula Weight 193.16
Percent Purity ≥98%
Chemical Name or Material trans-4-Nitrocinnamic acid
Hazard Category H315-H319-H335-H341
Hazard Statement GHS H Statement
H341-H315-H319-H335
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P261-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P501c
EINECSNumber 212-924-3
RTECSNumber GE0353000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only