missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans-4-Methylcyclohexylamine 98.0+%, TCI America™
Supplier: TCI America M17805G
Specifications
| trans-4-Methylcyclohexylamine | |
| Yellow | |
| C7H15N | |
| 5 g | |
| 4-methylcyclohexylamine, trans-4-methylcyclohexylamine, 4-methylcyclohexanamine, trans-4-methylcyclohexanamine, cyclohexanamine, 4-methyl, cis-4-methylcyclohexylamine, p-methylcyclohexylamine, trans-4-methyl-cyclohexylamine, trans-4-methylcyclohexyl amine, 4-methyl-cyclohexylamine | |
| CC1CCC(CC1)N | |
| 113.204 | |
| 113.20 | |
| Liquid |
| 2523-55-9 | |
| 151°C | |
| MFCD06411232 | |
| 2734 | |
| KSMVBYPXNKCPAJ-UHFFFAOYSA-N | |
| 4-methylcyclohexan-1-amine | |
| 80604 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 2523-55-9 | |
| 113.204 | |
| KSMVBYPXNKCPAJ-UHFFFAOYSA-N | |
| 80604 | |
| CC1CCC(CC1)N |
| C7H15N | |
| MFCD06411232 | |
| 4-methylcyclohexylamine, trans-4-methylcyclohexylamine, 4-methylcyclohexanamine, trans-4-methylcyclohexanamine, cyclohexanamine, 4-methyl, cis-4-methylcyclohexylamine, p-methylcyclohexylamine, trans-4-methyl-cyclohexylamine, trans-4-methylcyclohexyl amine, 4-methyl-cyclohexylamine | |
| 4-methylcyclohexan-1-amine |
Safety and Handling
TSCA : No