missing translation for 'onlineSavingsMsg'
Learn More

trans-4-Methylcyclohexylamine 98.0+%, TCI America™

Catalog No. p-7132851
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M17805G 5 g
M178025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M17805G Supplier TCI America Supplier No. M17805G

Chemical Identifiers

CAS 2523-55-9
Molecular Formula C7H15N
Molecular Weight (g/mol) 113.204
MDL Number MFCD06411232
InChI Key KSMVBYPXNKCPAJ-UHFFFAOYSA-N
Synonym 4-methylcyclohexylamine, trans-4-methylcyclohexylamine, 4-methylcyclohexanamine, trans-4-methylcyclohexanamine, cyclohexanamine, 4-methyl, cis-4-methylcyclohexylamine, p-methylcyclohexylamine, trans-4-methyl-cyclohexylamine, trans-4-methylcyclohexyl amine, 4-methyl-cyclohexylamine
PubChem CID 80604
IUPAC Name 4-methylcyclohexan-1-amine
SMILES CC1CCC(CC1)N

Specifications

Color Yellow
Boiling Point 151°C
Quantity 5 g
UN Number 2734
Formula Weight 113.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material trans-4-Methylcyclohexylamine