Learn More
trans-4-Methoxycinnamic acid, 98%
CAS: 943-89-5 | C10H10O3 | 178.19 g/mol
Supplier: Thermo Scientific Chemicals A1387009
Description
Esters derived from trans-4-methoxycinnamic acid are effective absorbers of UV radiation. The starting material was trans-4-methoxycinnamic acid and 3,5dimethoxybenzoic acid methyl ester in isolated avenanthramide alkaloids synthsis. trans-4-Methoxycinnamic acid was converted into its acid chloride (yield 98%) with thionyl chloride. Employed in organic synthesis of pharmaceutical intermediates, synthetic anti-adrenergic drugs esmolol, cosmetics ultraviolet absorption.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsEsters derived from trans-4-methoxycinnamic acid are effective absorbers of UV radiation. The starting material was trans-4-methoxycinnamic acid and 3,5dimethoxybenzoic acid methyl ester in isolated avenanthramide alkaloids synthsis. trans-4-Methoxycinnamic acid was converted into its acid chloride (yield 98%) with thionyl chloride. Employed in organic synthesis of pharmaceutical intermediates, synthetic anti-adrenergic drugs esmolol, cosmetics ultraviolet absorption.
Solubility
Soluble in ethanol, ethyl acetate and other organic solvents.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
| trans-4-Methoxycinnamic acid | |
| 171°C to 175°C | |
| MFCD00004398 | |
| 510468 | |
| Soluble in ethanol,ethyl acetate and other organic solvents. | |
| COC1=CC=C(\C=C\C(O)=O)C=C1 | |
| 178.19 | |
| 178.19 |
| 943-89-5 | |
| C10H10O3 | |
| 10 g | |
| 4-methoxycinnamic acid, p-methoxycinnamic acid, 3-4-methoxyphenyl acrylic acid, trans-4-methoxycinnamic acid, 4-methoxycinnamate, para-methoxycinnamic acid, o-methyl-p-coumaric acid, cinnamic acid, p-methoxy, e-3-4-methoxyphenyl acrylic acid, e-3-4-methoxyphenyl-2-propenoic acid | |
| AFDXODALSZRGIH-QPJJXVBHSA-N | |
| (E)-3-(4-methoxyphenyl)prop-2-enoic acid | |
| 699414 | |
| 98% |
Chemical Identifiers
| 943-89-5 | |
| 178.19 | |
| AFDXODALSZRGIH-QPJJXVBHSA-N | |
| 699414 | |
| COC1=CC=C(\C=C\C(O)=O)C=C1 |
| C10H10O3 | |
| MFCD00004398 | |
| 4-methoxycinnamic acid, p-methoxycinnamic acid, 3-4-methoxyphenyl acrylic acid, trans-4-methoxycinnamic acid, 4-methoxycinnamate, para-methoxycinnamic acid, o-methyl-p-coumaric acid, cinnamic acid, p-methoxy, e-3-4-methoxyphenyl acrylic acid, e-3-4-methoxyphenyl-2-propenoic acid | |
| (E)-3-(4-methoxyphenyl)prop-2-enoic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 213-405-4
RTECSNumber : UD3391300
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only