CAS RN 943-89-5
IUPAC Name:
(2E)-3-(4-methoxyphenyl)prop-2-enoic acid
Synonyms:
p-methoxy-cinnamic acid cinnamic acid, P-methoxy- P-methoxycinnamic acid (trans) methoxycinnamic acid
Molecular Weight (g/mol):
178.19
Molecular Formula:
C10H10O3
InChi Key:
AFDXODALSZRGIH-QPJJXVBHSA-N
SMILES:
COC1=CC=C(\C=C\C(O)=O)C=C1
1
–
1
of
1
results
1
Promotions Available
trans-4-Methoxycinnamic acid, 98%
trans-4-Methoxycinnamic acid, 98%
CAS: 943-89-5 Molecular Formula: C10H10O3 Molecular Weight (g/mol): 178.19 MDL Number: MFCD00004398 InChI Key: AFDXODALSZRGIH-QPJJXVBHSA-N Synonym: 4-methoxycinnamic acid,p-methoxycinnamic acid,3-4-methoxyphenyl acrylic acid,trans-4-methoxycinnamic acid,4-methoxycinnamate,para-methoxycinnamic acid,o-methyl-p-coumaric acid,cinnamic acid, p-methoxy,e-3-4-methoxyphenyl acrylic acid,e-3-4-methoxyphenyl-2-propenoic acid PubChem CID: 699414 IUPAC Name: (E)-3-(4-methoxyphenyl)prop-2-enoic acid SMILES: COC1=CC=C(\C=C\C(O)=O)C=C1