Learn More
trans-4-Methoxy-beta-nitrostyrene, 98%
CAS: 5576-97-6 | C9H9NO3 | 179.18 g/mol
Supplier: Thermo Scientific Chemicals A1274506
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| trans-4-Methoxy-beta-nitrostyrene | |
| 86°C to 88°C | |
| MFCD00024826 | |
| 2047935 | |
| JKQUXSHVQGBODD-UHFFFAOYSA-N | |
| 1-methoxy-4-[(E)-2-nitroethenyl]benzene | |
| 697963 | |
| 98% |
| 5576-97-6 | |
| C9H9NO3 | |
| 5 g | |
| 1-methoxy-4-2-nitrovinyl benzene, 4-methoxy-beta-nitrostyrene, p-2-nitrovinyl anisole, trans-4-methoxy-beta-nitrostyrene, e-1-methoxy-4-2-nitrovinyl benzene, 2'-nitro vinyl anisole, 1-methoxy-4-e-2-nitroethenyl benzene, 1-4-methoxyphenyl-2-nitroethene, 1-4-methoxyphenyl-2-nitroethylene, 4-2-nitrovinyl anisole | |
| COC1=CC=C(C=C[N+]([O-])=O)C=C1 | |
| 179.18 | |
| 179.18 |
Chemical Identifiers
| 5576-97-6 | |
| 179.18 | |
| JKQUXSHVQGBODD-UHFFFAOYSA-N | |
| 697963 |
| C9H9NO3 | |
| MFCD00024826 | |
| 1-methoxy-4-2-nitrovinyl benzene, 4-methoxy-beta-nitrostyrene, p-2-nitrovinyl anisole, trans-4-methoxy-beta-nitrostyrene, e-1-methoxy-4-2-nitrovinyl benzene, 2'-nitro vinyl anisole, 1-methoxy-4-e-2-nitroethenyl benzene, 1-4-methoxyphenyl-2-nitroethene, 1-4-methoxyphenyl-2-nitroethylene, 4-2-nitrovinyl anisole | |
| COC1=CC=C(C=C[N+]([O-])=O)C=C1 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only