missing translation for 'onlineSavingsMsg'
Learn More

trans-4-Hydroxy-L-proline methyl ester hydrochloride, 98%

Catalog No. p-7034078
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5172606 5 g
AAH5172614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5172606 Supplier Thermo Scientific Chemicals Supplier No. H5172606

CAS: 40216-83-9 | C6H12ClNO3 | 181.616 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 40216-83-9
Molecular Formula C6H12ClNO3
Molecular Weight (g/mol) 181.616
MDL Number MFCD00080855
InChI Key KLGSHNXEUZOKHH-JBUOLDKXSA-N
Synonym 2s,4r-methyl 4-hydroxypyrrolidine-2-carboxylate hydrochloride, trans-4-hydroxy-l-proline methyl ester hydrochloride, h-hyp-ome.hcl, methyl 2s,4r-4-hydroxypyrrolidine-2-carboxylate hydrochloride, l-4-hydroxyproline methyl ester hydrochloride, hydroxyproline methyl ester hydrochloride, trans-4-hydroxy-l-proline methyl ester hcl, 2s,4r-methyl 4-hydroxypyrrolidine-2-carboxylate hcl, trans-4-hydroxy-l-proline methyl ester, hydrochloride salt, l-proline, 4-hydroxy-, methyl ester, hydrochloride, 4r
PubChem CID 11435319
IUPAC Name methyl (2S,4R)-4-hydroxypyrrolidine-2-carboxylate;hydrochloride
SMILES COC(=O)C1CC(CN1)O.Cl

Specifications

Beilstein 4716932
Quantity 5 g
Formula Weight 181.62
Percent Purity 98%
Chemical Name or Material trans-4-Hydroxy-L-proline methyl ester hydrochloride
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only