accessibility menu, dialog, popup

UserName

ClCH3OO(S)(R)OHHNH

CAS RN 40216-83-9

IUPAC Name: hydrogen methyl (2S,4R)-4-hydroxypyrrolidine-2-carboxylate chloride
Synonyms: hydrogen methyl (2S,4R)-4-hydroxypyrrolidine-2-carboxylate chloride
Molecular Weight (g/mol): 181.62
Molecular Formula: C6H12ClNO3
InChi Key: KLGSHNXEUZOKHH-JBUOLDKXSA-N
SMILES: [H+].[Cl-].COC(=O)[C@@H]1C[C@@H](O)CN1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)