missing translation for 'onlineSavingsMsg'
Learn More

trans-4-Hydroxy-L-proline, ≥99% (TLC), MilliporeSigma™ Supelco™

Catalog No. 111014402
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014402 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014402 Supplier Merck Emd Millipore Supplier No. 41875100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
CAS 51-35-4
Physical Form Neat
Linear Formula C5H9 NO3
MDL Number MFCD00064320
Melting Point 273°C
Synonym (2 S,4 R)-4-Hydroxypyrrolidine-2-carboxylic acid; Hyp
Type Analytical Standard
Quantity 100 mg
Formula Weight 131.13
Percent Purity ≥99% (TLC)
Grade Analytical Standard
Show More Show Less