missing translation for 'onlineSavingsMsg'
Learn More

trans-4-Hydroxy-D-proline, 97%

CAS: 3398-22-9 | C5H9NO3 | 131.13 g/mol

Supplier:  Thermo Scientific Chemicals H2742103

 View more versions of this product

Catalog No. AAH2742103


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

∽260°C (decomposition)
3398-22-9
MFCD00065608
PMMYEEVYMWASQN-UHFFFAOYNA-N
(2R,4S)-4-hydroxypyrrolidine-2-carboxylic acid
131.13
CHEBI:27992
97%
trans-4-Hydroxy-D-proline
C5H9NO3
trans-4-hydroxy-d-proline, 2r,4s-4-hydroxypyrrolidine-2-carboxylic acid, h-d-hyp-oh, d-proline, 4-hydroxy-, 4s, 2r,4s-4-hydroxy-pyrrolidine-2-carboxylic acid, d-hydroxyproline, ambotzhaa1169, pubchem9966, d-hyp-oh
OC1CNC(C1)C(O)=O
1 g
440074
131.13

Chemical Identifiers

3398-22-9
131.13
PMMYEEVYMWASQN-UHFFFAOYNA-N
440074
(2R,4S)-4-hydroxypyrrolidine-2-carboxylic acid
C5H9NO3
MFCD00065608
trans-4-hydroxy-d-proline, 2r,4s-4-hydroxypyrrolidine-2-carboxylic acid, h-d-hyp-oh, d-proline, 4-hydroxy-, 4s, 2r,4s-4-hydroxy-pyrrolidine-2-carboxylic acid, d-hydroxyproline, ambotzhaa1169, pubchem9966, d-hyp-oh
CHEBI:27992
OC1CNC(C1)C(O)=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only