missing translation for 'onlineSavingsMsg'
Learn More

trans-4-Hydroxy-D-proline, 97%

Catalog No. p-7029902
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH27421MD 250 mg
AAH2742103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH27421MD Supplier Thermo Scientific Chemicals Supplier No. H27421MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3398-22-9 | C5H9NO3 | 131.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3398-22-9
Molecular Formula C5H9NO3
Molecular Weight (g/mol) 131.13
MDL Number MFCD00065608
InChI Key PMMYEEVYMWASQN-UHFFFAOYNA-N
Synonym trans-4-hydroxy-d-proline, 2r,4s-4-hydroxypyrrolidine-2-carboxylic acid, h-d-hyp-oh, d-proline, 4-hydroxy-, 4s, 2r,4s-4-hydroxy-pyrrolidine-2-carboxylic acid, d-hydroxyproline, ambotzhaa1169, pubchem9966, d-hyp-oh
PubChem CID 440074
ChEBI CHEBI:27992
IUPAC Name (2R,4S)-4-hydroxypyrrolidine-2-carboxylic acid
SMILES OC1CNC(C1)C(O)=O

Specifications

Melting Point ∽260°C (decomposition)
Quantity 250 mg
Formula Weight 131.13
Percent Purity 97%
Chemical Name or Material trans-4-Hydroxy-D-proline
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only