Learn More
trans-4-Hydroxy-3-methoxycinnamic acid, 99%
CAS: 537-98-4 | C10H10O4 | 194.19 g/mol
Supplier: Thermo Scientific Chemicals A1389036
Description
A chemopreventive analogue of caffeic acid.trans-4-Hydroxy-3-methoxycinnamic acid is used as an important raw material for the preparation of its corresponding esters by reacting with alcohols.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsA chemopreventive analogue of caffeic acid.trans-4-Hydroxy-3-methoxycinnamic acid is used as an important raw material for the preparation of its corresponding esters by reacting with alcohols.
Solubility
Soluble in alcohol and hot water.
Notes
Incompatible with oxidizing agents.
Specifications
| trans-4-Hydroxy-3-methoxycinnamic acid | |
| 170°C to 175°C | |
| MFCD00004400 | |
| 1371483 | |
| ferulic acid, trans-ferulic acid, 4-hydroxy-3-methoxycinnamic acid, trans-4-hydroxy-3-methoxycinnamic acid, 3-4-hydroxy-3-methoxyphenyl acrylic acid, e-ferulic acid, coniferic acid, ferulate, ferulic acid, trans, 2-propenoic acid, 3-4-hydroxy-3-methoxyphenyl | |
| KSEBMYQBYZTDHS-HWKANZROSA-N | |
| (E)-3-(4-hydroxy-3-methoxyphenyl)prop-2-enoic acid | |
| 445858 | |
| 194.19 |
| 537-98-4 | |
| C10H10O4 | |
| 500 g | |
| 14,4062 | |
| Soluble in alcohol and hot water. | |
| COC1=CC(\C=C\C(O)=O)=CC=C1O | |
| 194.19 | |
| CHEBI:17620 | |
| 99% |
Chemical Identifiers
| 537-98-4 | |
| 194.19 | |
| KSEBMYQBYZTDHS-HWKANZROSA-N | |
| 445858 | |
| (E)-3-(4-hydroxy-3-methoxyphenyl)prop-2-enoic acid |
| C10H10O4 | |
| MFCD00004400 | |
| ferulic acid, trans-ferulic acid, 4-hydroxy-3-methoxycinnamic acid, trans-4-hydroxy-3-methoxycinnamic acid, 3-4-hydroxy-3-methoxyphenyl acrylic acid, e-ferulic acid, coniferic acid, ferulate, ferulic acid, trans, 2-propenoic acid, 3-4-hydroxy-3-methoxyphenyl | |
| CHEBI:17620 | |
| COC1=CC(\C=C\C(O)=O)=CC=C1O |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 208-679-7
RTECSNumber : UD3365500
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only