missing translation for 'onlineSavingsMsg'
Learn More

trans-4-Hydroxy-3-methoxycinnamic acid, 99%

Catalog No. p-7021527
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1389014 25 g
AAA1389022 100 g
AAA1389036 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1389014 Supplier Thermo Scientific Chemicals Supplier No. A1389014
Only null left

CAS: 537-98-4 | C10H10O4 | 194.19 g/mol

A chemopreventive analogue of caffeic acid.trans-4-Hydroxy-3-methoxycinnamic acid is used as an important raw material for the preparation of its corresponding esters by reacting with alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A chemopreventive analogue of caffeic acid.trans-4-Hydroxy-3-methoxycinnamic acid is used as an important raw material for the preparation of its corresponding esters by reacting with alcohols.

Solubility
Soluble in alcohol and hot water.

Notes
Incompatible with oxidizing agents.

Chemical Identifiers

CAS 537-98-4
Molecular Formula C10H10O4
Molecular Weight (g/mol) 194.19
MDL Number MFCD00004400
InChI Key KSEBMYQBYZTDHS-HWKANZROSA-N
Synonym ferulic acid, trans-ferulic acid, 4-hydroxy-3-methoxycinnamic acid, trans-4-hydroxy-3-methoxycinnamic acid, 3-4-hydroxy-3-methoxyphenyl acrylic acid, e-ferulic acid, coniferic acid, ferulate, ferulic acid, trans, 2-propenoic acid, 3-4-hydroxy-3-methoxyphenyl
PubChem CID 445858
ChEBI CHEBI:17620
IUPAC Name (E)-3-(4-hydroxy-3-methoxyphenyl)prop-2-enoic acid
SMILES COC1=CC(\C=C\C(O)=O)=CC=C1O

Specifications

Melting Point 170°C to 175°C
Quantity 25 g
Beilstein 1371483
Merck Index 14,4062
Solubility Information Soluble in alcohol and hot water.
Formula Weight 194.19
Percent Purity 99%
Chemical Name or Material trans-4-Hydroxy-3-methoxycinnamic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 208-679-7
RTECSNumber UD3365500
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only