missing translation for 'onlineSavingsMsg'
Learn More

trans-4-Hydroxy-3-methoxycinnamic acid, 99%

CAS: 537-98-4 | C10H10O4 | 194.19 g/mol

Supplier:  Thermo Scientific Chemicals A1389022

 View more versions of this product

Catalog No. AAA1389022


Only null left
Explore available promotions

Description

Description

A chemopreventive analogue of caffeic acid.trans-4-Hydroxy-3-methoxycinnamic acid is used as an important raw material for the preparation of its corresponding esters by reacting with alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A chemopreventive analogue of caffeic acid.trans-4-Hydroxy-3-methoxycinnamic acid is used as an important raw material for the preparation of its corresponding esters by reacting with alcohols.

Solubility
Soluble in alcohol and hot water.

Notes
Incompatible with oxidizing agents.
Specifications

Specifications

trans-4-Hydroxy-3-methoxycinnamic acid
170°C to 175°C
MFCD00004400
1371483
ferulic acid, trans-ferulic acid, 4-hydroxy-3-methoxycinnamic acid, trans-4-hydroxy-3-methoxycinnamic acid, 3-4-hydroxy-3-methoxyphenyl acrylic acid, e-ferulic acid, coniferic acid, ferulate, ferulic acid, trans, 2-propenoic acid, 3-4-hydroxy-3-methoxyphenyl
KSEBMYQBYZTDHS-HWKANZROSA-N
(E)-3-(4-hydroxy-3-methoxyphenyl)prop-2-enoic acid
445858
194.19
537-98-4
C10H10O4
100 g
14,4062
Soluble in alcohol and hot water.
COC1=CC(\C=C\C(O)=O)=CC=C1O
194.19
CHEBI:17620
99%

Chemical Identifiers

537-98-4
194.19
KSEBMYQBYZTDHS-HWKANZROSA-N
445858
(E)-3-(4-hydroxy-3-methoxyphenyl)prop-2-enoic acid
C10H10O4
MFCD00004400
ferulic acid, trans-ferulic acid, 4-hydroxy-3-methoxycinnamic acid, trans-4-hydroxy-3-methoxycinnamic acid, 3-4-hydroxy-3-methoxyphenyl acrylic acid, e-ferulic acid, coniferic acid, ferulate, ferulic acid, trans, 2-propenoic acid, 3-4-hydroxy-3-methoxyphenyl
CHEBI:17620
COC1=CC(\C=C\C(O)=O)=CC=C1O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 208-679-7

RTECSNumber : UD3365500

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only