missing translation for 'onlineSavingsMsg'
Learn More

trans-4-Fluoro-L-proline 98.0+%, TCI America™

Catalog No. F081850MG
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F081850MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F081850MG Supplier TCI America Supplier No. F081850MG

Chemical Identifiers

CAS 2507-61-1
Molecular Formula C5H8FNO2
Molecular Weight (g/mol) 133.122
MDL Number MFCD02683982
InChI Key ZIWHMENIDGOELV-DMTCNVIQSA-N
Synonym (2S,4R)-4-Fluoropyrrolidine-2-carboxylic Acid
PubChem CID 11902999
IUPAC Name (2S,4R)-4-fluoropyrrolidine-2-carboxylic acid
SMILES C1C(CNC1C(=O)O)F

Specifications

Melting Point 264°C
Color Yellow
Quantity 50 mg
Formula Weight 133.12
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material trans-4-Fluoro-L-proline