accessibility menu, dialog, popup

UserName

HNOOHF

CAS RN 2507-61-1

IUPAC Name: 4-fluoropyrrolidine-2-carboxylic acid
Synonyms: 4-fluoropyrrolidine-2-carboxylic acid
Molecular Weight (g/mol): 133.12
Molecular Formula: C5H8FNO2
InChi Key: ZIWHMENIDGOELV-UHFFFAOYNA-N
SMILES: OC(=O)C1CC(F)CN1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results