missing translation for 'onlineSavingsMsg'
Learn More

trans-4-(Carbobenzoxyamino)cyclohexanecarboxylic Acid 96.0+%, TCI America™

Catalog No. C20361G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C20361G 1 g
C20365G 5 g
C203625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C20361G Supplier TCI America Supplier No. C20361G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 34771-04-5
Molecular Formula C15H19NO4
Molecular Weight (g/mol) 277.32
InChI Key ZVMICQYOGWAOSU-UHFFFAOYSA-N
Synonym trans-4-carbobenzoxyamino cyclohexanecarboxylic acid, trans-4-cbz-amino cyclohexanecarboxylic acid, 4-benzyloxycarbonylamino-cyclohexanecarboxylic acid, trans-4-benzyloxycarbonylaminocyclohexanecarboxylic acid, cis-4-benzyloxycarbonylaminocyclohexanecarboxylic acid, cis-4-benzyloxycarbonylaminocyclohexane-carboxylic acid, 4-benzyloxy carbonyl amino cyclohexanecarboxylic acid, 4-phenylmethoxycarbonylamino cyclohexane-1-carboxylic acid, trans-4-benzyloxy carbonyl amino cyclohexanecarboxylic acid
PubChem CID 285247
IUPAC Name 4-(phenylmethoxycarbonylamino)cyclohexane-1-carboxylic acid
SMILES C1CC(CCC1C(=O)O)NC(=O)OCC2=CC=CC=C2

Specifications

Melting Point 217°C
Color White
Quantity 1 g
Formula Weight 277.32
Percent Purity ≥96.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material trans-4-(Carbobenzoxyamino)cyclohexanecarboxylic Acid