accessibility menu, dialog, popup

UserName

OHONHOO

CAS RN 34771-04-5

IUPAC Name: 4-{[(benzyloxy)carbonyl]amino}cyclohexane-1-carboxylic acid
Synonyms: 4-{[(benzyloxy)carbonyl]amino}cyclohexane-1-carboxylic acid
Molecular Weight (g/mol): 277.32
Molecular Formula: C15H19NO4
InChi Key: ZVMICQYOGWAOSU-UHFFFAOYSA-N
SMILES: OC(=O)C1CCC(CC1)NC(=O)OCC1=CC=CC=C1
Percent Purity 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)