missing translation for 'onlineSavingsMsg'
Learn More

trans-4-Bromostilbene 98.0+%, TCI America™

Catalog No. B24165G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B24165G 5 g
B24161G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B24165G Supplier TCI America Supplier No. B24165G

Chemical Identifiers

CAS 13041-70-8
Molecular Formula C14H11Br
Molecular Weight (g/mol) 259.146
MDL Number MFCD00155020
InChI Key ZZMMKLVIBZWGPK-VOTSOKGWSA-N
Synonym 4-bromostilbene, 1-bromo-4-styrylbenzene, e-1-bromo-4-styrylbenzene, 4-bromo-trans-stilbene, e-4-bromostilbene, 1-bromo-4-e-2-phenylethenyl benzene, z-4-bromostilbene, benzene, 1-bromo-4-1e-2-phenylethenyl, benzene, 1-bromo-4-2-phenylethenyl
PubChem CID 5374623
IUPAC Name 1-bromo-4-[(E)-2-phenylethenyl]benzene
SMILES C1=CC=C(C=C1)C=CC2=CC=C(C=C2)Br

Specifications

Melting Point 140°C
Color White
Quantity 5 g
Formula Weight 259.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material trans-4-Bromostilbene