missing translation for 'onlineSavingsMsg'
Learn More

trans-4-(Boc-amino)cyclohexaneacetic acid, 97%

Catalog No. AAH6205806
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6205806 5 g
AAH6205803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6205806 Supplier Thermo Scientific Chemicals Supplier No. H6205806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 189153-10-4 | C13H23NO4 | 257.33 g/mol

trans-4-(Boc-amino)cyclohexaneacetic acid used as a intermediate in pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-4-(Boc-amino)cyclohexaneacetic acid used as a intermediate in pharmaceutical.

Solubility
Slightly soluble in water (1.1 g/L) (25°C)

Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away from oxidizing agent.

Chemical Identifiers

CAS 189153-10-4
Molecular Formula C13H23NO4
Molecular Weight (g/mol) 257.33
MDL Number MFCD03844588
InChI Key IHXBNSUFUFFBRL-UHFFFAOYSA-N
Synonym 2-cis-4-tert-butoxycarbonyl amino cyclohexyl acetic acid, trans-n-boc-4-aminocyclohexyl acetic acid, 2-trans-4-tert-butoxycarbonyl amino cyclohexyl acetic acid, boc-1,4-trans-acha-oh, cis-4-boc-amino cyclohexyl acetic acid, trans-n-boc-4-aminocyclohexyl aceticacid, 4-tert-butoxycarbonylamino-cyclohexyl-acetic acid, trans-4-tert-butoxycarbonylamino cyclohexylacetic acid, 1r,4r-4-tert-butoxycarbonyl amino cyclohexyl acetic acid, 1s,4s-4-tert-butoxycarbonyl amino cyclohexyl acetic acid
PubChem CID 11253897
IUPAC Name 2-[4-[(2-methylpropan-2-yl)oxycarbonylamino]cyclohexyl]acetic acid
SMILES CC(C)(C)OC(=O)NC1CCC(CC1)CC(=O)O

Specifications

Solubility Information Slightly soluble in water (1.1g/L) (25°C)
Quantity 5 g
Formula Weight 257.32
Percent Purity 97%
Chemical Name or Material trans-4-(Boc-amino)cyclohexaneacetic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only