accessibility menu, dialog, popup

UserName

H3CH3CCH3OONHOHO

CAS RN 189153-10-4

IUPAC Name: 2-(4-{[(tert-butoxy)carbonyl]amino}cyclohexyl)acetic acid
Synonyms: {4-[(tert-butoxycarbonyl)amino]cyclohexyl}acetic acid
Molecular Weight (g/mol): 257.33
Molecular Formula: C13H23NO4
InChi Key: IHXBNSUFUFFBRL-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NC1CCC(CC(O)=O)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)