Learn More
trans-4-(Benzyloxycarbonylamino)cyclohexanemethylamine, 97%, Thermo Scientific Chemicals
$1512.89 - $1512.89
Chemical Identifiers
| CAS | 177582-74-0 |
|---|---|
| Molecular Formula | C15H22N2O2 |
| Molecular Weight (g/mol) | 262.35 |
| MDL Number | MFCD06657669 |
| InChI Key | AJYOPGOCTHCBTL-UHFFFAOYSA-N |
| Synonym | benzyl trans-4-aminomethyl cyclohexyl carbamate, benzyl trans-4-aminomethylcyclohexylcarbamate, benzyl 4-aminomethyl cyclohexyl carbamate, benzyl n-1r,4r-4-aminomethyl cyclohexyl carbamate, ajyopgocthcbtl-mqmhxkeqsa-n, ajyopgocthcbtl-xbxgtlagsa-n, benzyl 4-aminomethyl cyclohexylcarbamate, benzyl n-4-aminomethyl cyclohexyl carbamate, benzyl cis-4-aminomethyl cyclohexyl carbamate, benzyl trans-4-aminomethylcyclohexyl carbamate |
| PubChem CID | 22253963 |
| IUPAC Name | benzyl N-[4-(aminomethyl)cyclohexyl]carbamate |
| SMILES | NCC1CCC(CC1)NC(=O)OCC1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH6203406
|
Thermo Scientific Chemicals
H6203406 |
5 g |
Each for $1,512.89
|
|
|||||
Description
trans-4-(Benzyloxycarbonylamino)cyclohexanemethylamine used as intermediate for chemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applicationstrans-4-(Benzyloxycarbonylamino)cyclohexanemethylamine used as intermediate for chemical research.
Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away from oxidizing agent.
Chemical Identifiers
| 177582-74-0 | |
| 262.35 | |
| AJYOPGOCTHCBTL-UHFFFAOYSA-N | |
| 22253963 | |
| NCC1CCC(CC1)NC(=O)OCC1=CC=CC=C1 |
| C15H22N2O2 | |
| MFCD06657669 | |
| benzyl trans-4-aminomethyl cyclohexyl carbamate, benzyl trans-4-aminomethylcyclohexylcarbamate, benzyl 4-aminomethyl cyclohexyl carbamate, benzyl n-1r,4r-4-aminomethyl cyclohexyl carbamate, ajyopgocthcbtl-mqmhxkeqsa-n, ajyopgocthcbtl-xbxgtlagsa-n, benzyl 4-aminomethyl cyclohexylcarbamate, benzyl n-4-aminomethyl cyclohexyl carbamate, benzyl cis-4-aminomethyl cyclohexyl carbamate, benzyl trans-4-aminomethylcyclohexyl carbamate | |
| benzyl N-[4-(aminomethyl)cyclohexyl]carbamate |
Specifications
| 177582-74-0 | |
| MFCD06657669 | |
| benzyl trans-4-aminomethyl cyclohexyl carbamate, benzyl trans-4-aminomethylcyclohexylcarbamate, benzyl 4-aminomethyl cyclohexyl carbamate, benzyl n-1r,4r-4-aminomethyl cyclohexyl carbamate, ajyopgocthcbtl-mqmhxkeqsa-n, ajyopgocthcbtl-xbxgtlagsa-n, benzyl 4-aminomethyl cyclohexylcarbamate, benzyl n-4-aminomethyl cyclohexyl carbamate, benzyl cis-4-aminomethyl cyclohexyl carbamate, benzyl trans-4-aminomethylcyclohexyl carbamate | |
| NCC1CCC(CC1)NC(=O)OCC1=CC=CC=C1 | |
| 5 g | |
| 22253963 | |
| 97% |
| C15H22N2O2 | |
| 7710506 | |
| AJYOPGOCTHCBTL-UHFFFAOYSA-N | |
| benzyl N-[4-(aminomethyl)cyclohexyl]carbamate | |
| 262.35 | |
| 262.34 | |
| trans-4-(Benzyloxycarbonylamino)cyclohexanemethylamine |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only