missing translation for 'onlineSavingsMsg'
Learn More

trans-4-(Benzyloxycarbonylamino)cyclohexanemethylamine, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H6203406

 View more versions of this product

Catalog No. AAH6203406


Only null left

Description

Description

trans-4-(Benzyloxycarbonylamino)cyclohexanemethylamine used as intermediate for chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-4-(Benzyloxycarbonylamino)cyclohexanemethylamine used as intermediate for chemical research.

Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away from oxidizing agent.
Specifications

Specifications

trans-4-(Benzyloxycarbonylamino)cyclohexanemethylamine
C15H22N2O2
7710506
AJYOPGOCTHCBTL-UHFFFAOYSA-N
benzyl N-[4-(aminomethyl)cyclohexyl]carbamate
262.35
262.34
177582-74-0
MFCD06657669
benzyl trans-4-aminomethyl cyclohexyl carbamate, benzyl trans-4-aminomethylcyclohexylcarbamate, benzyl 4-aminomethyl cyclohexyl carbamate, benzyl n-1r,4r-4-aminomethyl cyclohexyl carbamate, ajyopgocthcbtl-mqmhxkeqsa-n, ajyopgocthcbtl-xbxgtlagsa-n, benzyl 4-aminomethyl cyclohexylcarbamate, benzyl n-4-aminomethyl cyclohexyl carbamate, benzyl cis-4-aminomethyl cyclohexyl carbamate, benzyl trans-4-aminomethylcyclohexyl carbamate
NCC1CCC(CC1)NC(=O)OCC1=CC=CC=C1
5 g
22253963
97%

Chemical Identifiers

177582-74-0
262.35
AJYOPGOCTHCBTL-UHFFFAOYSA-N
22253963
NCC1CCC(CC1)NC(=O)OCC1=CC=CC=C1
C15H22N2O2
MFCD06657669
benzyl trans-4-aminomethyl cyclohexyl carbamate, benzyl trans-4-aminomethylcyclohexylcarbamate, benzyl 4-aminomethyl cyclohexyl carbamate, benzyl n-1r,4r-4-aminomethyl cyclohexyl carbamate, ajyopgocthcbtl-mqmhxkeqsa-n, ajyopgocthcbtl-xbxgtlagsa-n, benzyl 4-aminomethyl cyclohexylcarbamate, benzyl n-4-aminomethyl cyclohexyl carbamate, benzyl cis-4-aminomethyl cyclohexyl carbamate, benzyl trans-4-aminomethylcyclohexyl carbamate
benzyl N-[4-(aminomethyl)cyclohexyl]carbamate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only