missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
trans-4-Amylcyclohexanol 93.0+%, TCI America™
Supplier: TCI America A23151G
Specifications
trans-4-Amylcyclohexanol | |
Yellow | |
MFCD01017412,MFCD11114412 | |
trans-4-Pentylcyclohexanol | |
CCCCCC1CCC(O)CC1 | |
170.30 | |
170.30 | |
Liquid |
77866-59-2 | |
C11H22O | |
1 g | |
VHWGPISIUNUREA-UHFFFAOYSA-N | |
4-pentylcyclohexan-1-ol | |
41076 | |
≥93.0% (GC) |
Chemical Identifiers
77866-59-2 | |
170.30 | |
VHWGPISIUNUREA-UHFFFAOYSA-N | |
41076 | |
CCCCCC1CCC(O)CC1 |
C11H22O | |
MFCD01017412,MFCD11114412 | |
trans-4-Pentylcyclohexanol | |
4-pentylcyclohexan-1-ol |
Safety and Handling
TSCA : No