accessibility menu, dialog, popup

UserName

CAS RN 77866-59-2

H3COH

CAS RN 77866-59-2

IUPAC Name: 4-pentylcyclohexan-1-ol
Synonyms: 4-pentylcyclohexan-1-ol
Molecular Weight (g/mol): 170.3
Molecular Formula: C11H22O
InChi Key: VHWGPISIUNUREA-UHFFFAOYSA-N
SMILES: CCCCCC1CCC(O)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results