missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans-4-Aminocyclohexanecarboxylic Acid 98.0+%, TCI America™
Supplier: TCI America A165625G
Specifications
| 495°C | |
| 3685-25-4 | |
| C7H13NO2 | |
| DRNGLYHKYPNTEA-UHFFFAOYSA-N | |
| 4-aminocyclohexane-1-carboxylic acid | |
| 143.186 | |
| 143.19 | |
| Crystalline Powder |
| trans-4-Aminocyclohexanecarboxylic Acid | |
| White | |
| 4-aminocyclohexanecarboxylic acid, trans-4-aminocyclohexanecarboxylic acid, cis-4-aminocyclohexanecarboxylic acid, cis-4-aminocyclohexane carboxylic acid, cis-4-amino-1-cyclohexanecarboxylic acid, trans-4-amino-cyclohexanecarboxylic acid, cis-4-aminocyclohexanecarboxylicacid, cyclohexanecarboxylic acid, 4-amino-, trans, trans-4-aminocyclohexane carboxylic acid, 1s,4s-4-aminocyclohexanecarboxylic acid | |
| C1CC(CCC1C(=O)O)N | |
| 25 g | |
| 171560 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 3685-25-4 | |
| 143.186 | |
| 4-aminocyclohexanecarboxylic acid, trans-4-aminocyclohexanecarboxylic acid, cis-4-aminocyclohexanecarboxylic acid, cis-4-aminocyclohexane carboxylic acid, cis-4-amino-1-cyclohexanecarboxylic acid, trans-4-amino-cyclohexanecarboxylic acid, cis-4-aminocyclohexanecarboxylicacid, cyclohexanecarboxylic acid, 4-amino-, trans, trans-4-aminocyclohexane carboxylic acid, 1s,4s-4-aminocyclohexanecarboxylic acid | |
| 4-aminocyclohexane-1-carboxylic acid |
| C7H13NO2 | |
| DRNGLYHKYPNTEA-UHFFFAOYSA-N | |
| 171560 | |
| C1CC(CCC1C(=O)O)N |
Safety and Handling
TSCA : No