missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans-4-Aminocyclohexanecarboxylic Acid 98.0+%, TCI America™
$100.11 - $1133.93
Chemical Identifiers
| CAS | 3685-25-4 |
|---|---|
| Molecular Formula | C7H13NO2 |
| Molecular Weight (g/mol) | 143.186 |
| InChI Key | DRNGLYHKYPNTEA-UHFFFAOYSA-N |
| Synonym | 4-aminocyclohexanecarboxylic acid, trans-4-aminocyclohexanecarboxylic acid, cis-4-aminocyclohexanecarboxylic acid, cis-4-aminocyclohexane carboxylic acid, cis-4-amino-1-cyclohexanecarboxylic acid, trans-4-amino-cyclohexanecarboxylic acid, cis-4-aminocyclohexanecarboxylicacid, cyclohexanecarboxylic acid, 4-amino-, trans, trans-4-aminocyclohexane carboxylic acid, 1s,4s-4-aminocyclohexanecarboxylic acid |
| PubChem CID | 171560 |
| IUPAC Name | 4-aminocyclohexane-1-carboxylic acid |
| SMILES | C1CC(CCC1C(=O)O)N |
Chemical Identifiers
| 3685-25-4 | |
| 143.186 | |
| 4-aminocyclohexanecarboxylic acid, trans-4-aminocyclohexanecarboxylic acid, cis-4-aminocyclohexanecarboxylic acid, cis-4-aminocyclohexane carboxylic acid, cis-4-amino-1-cyclohexanecarboxylic acid, trans-4-amino-cyclohexanecarboxylic acid, cis-4-aminocyclohexanecarboxylicacid, cyclohexanecarboxylic acid, 4-amino-, trans, trans-4-aminocyclohexane carboxylic acid, 1s,4s-4-aminocyclohexanecarboxylic acid | |
| 4-aminocyclohexane-1-carboxylic acid |
| C7H13NO2 | |
| DRNGLYHKYPNTEA-UHFFFAOYSA-N | |
| 171560 | |
| C1CC(CCC1C(=O)O)N |
Specifications
| 495°C | |
| White | |
| 4-aminocyclohexanecarboxylic acid, trans-4-aminocyclohexanecarboxylic acid, cis-4-aminocyclohexanecarboxylic acid, cis-4-aminocyclohexane carboxylic acid, cis-4-amino-1-cyclohexanecarboxylic acid, trans-4-amino-cyclohexanecarboxylic acid, cis-4-aminocyclohexanecarboxylicacid, cyclohexanecarboxylic acid, 4-amino-, trans, trans-4-aminocyclohexane carboxylic acid, 1s,4s-4-aminocyclohexanecarboxylic acid | |
| C1CC(CCC1C(=O)O)N | |
| 1 g | |
| 171560 | |
| ≥98.0% (GC,T) | |
| trans-4-Aminocyclohexanecarboxylic Acid |
| 3685-25-4 | |
| C7H13NO2 | |
| DRNGLYHKYPNTEA-UHFFFAOYSA-N | |
| 4-aminocyclohexane-1-carboxylic acid | |
| 143.186 | |
| 143.19 | |
| Crystalline Powder |
Safety and Handling
TSCA : No