missing translation for 'onlineSavingsMsg'
Learn More

trans-4-Aminocyclohexanecarboxylic acid, 97%

CAS: 3685-25-4 | C7H13NO2 | 143.186 g/mol

$381.30 - $1156.27

Chemical Identifiers

CAS 3685-25-4
Molecular Formula C7H13NO2
Molecular Weight (g/mol) 143.186
MDL Number MFCD00209953
InChI Key DRNGLYHKYPNTEA-UHFFFAOYSA-N
Synonym 4-aminocyclohexanecarboxylic acid, trans-4-aminocyclohexanecarboxylic acid, cis-4-aminocyclohexanecarboxylic acid, cis-4-aminocyclohexane carboxylic acid, cis-4-amino-1-cyclohexanecarboxylic acid, trans-4-amino-cyclohexanecarboxylic acid, cis-4-aminocyclohexanecarboxylicacid, cyclohexanecarboxylic acid, 4-amino-, trans, trans-4-aminocyclohexane carboxylic acid, 1s,4s-4-aminocyclohexanecarboxylic acid
PubChem CID 171560
IUPAC Name 4-aminocyclohexane-1-carboxylic acid
SMILES C1CC(CCC1C(=O)O)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2748903
View Documents
Thermo Scientific Chemicals
H2748903
1 g
Each for $381.30
Only null left
 
AAH2748906
View Documents
Thermo Scientific Chemicals
H2748906
5 g
Each for $1,156.27
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

3685-25-4
143.186
DRNGLYHKYPNTEA-UHFFFAOYSA-N
171560
C1CC(CCC1C(=O)O)N
C7H13NO2
MFCD00209953
4-aminocyclohexanecarboxylic acid, trans-4-aminocyclohexanecarboxylic acid, cis-4-aminocyclohexanecarboxylic acid, cis-4-aminocyclohexane carboxylic acid, cis-4-amino-1-cyclohexanecarboxylic acid, trans-4-amino-cyclohexanecarboxylic acid, cis-4-aminocyclohexanecarboxylicacid, cyclohexanecarboxylic acid, 4-amino-, trans, trans-4-aminocyclohexane carboxylic acid, 1s,4s-4-aminocyclohexanecarboxylic acid
4-aminocyclohexane-1-carboxylic acid

Specifications

>300°C
C7H13NO2
4-aminocyclohexanecarboxylic acid, trans-4-aminocyclohexanecarboxylic acid, cis-4-aminocyclohexanecarboxylic acid, cis-4-aminocyclohexane carboxylic acid, cis-4-amino-1-cyclohexanecarboxylic acid, trans-4-amino-cyclohexanecarboxylic acid, cis-4-aminocyclohexanecarboxylicacid, cyclohexanecarboxylic acid, 4-amino-, trans, trans-4-aminocyclohexane carboxylic acid, 1s,4s-4-aminocyclohexanecarboxylic acid
C1CC(CCC1C(=O)O)N
1 g
171560
97%
3685-25-4
MFCD00209953
DRNGLYHKYPNTEA-UHFFFAOYSA-N
4-aminocyclohexane-1-carboxylic acid
143.186
143.19
trans-4-Aminocyclohexanecarboxylic acid
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only