missing translation for 'onlineSavingsMsg'
Learn More

trans-4-Aminocyclohexanecarboxylic acid, 97%

CAS: 3685-25-4 | C7H13NO2 | 143.186 g/mol

Supplier:  Thermo Scientific Chemicals H2748906

 View more versions of this product

Catalog No. AAH2748906


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

>300°C
3685-25-4
MFCD00209953
DRNGLYHKYPNTEA-UHFFFAOYSA-N
4-aminocyclohexane-1-carboxylic acid
143.186
143.19
trans-4-Aminocyclohexanecarboxylic acid
C7H13NO2
4-aminocyclohexanecarboxylic acid, trans-4-aminocyclohexanecarboxylic acid, cis-4-aminocyclohexanecarboxylic acid, cis-4-aminocyclohexane carboxylic acid, cis-4-amino-1-cyclohexanecarboxylic acid, trans-4-amino-cyclohexanecarboxylic acid, cis-4-aminocyclohexanecarboxylicacid, cyclohexanecarboxylic acid, 4-amino-, trans, trans-4-aminocyclohexane carboxylic acid, 1s,4s-4-aminocyclohexanecarboxylic acid
C1CC(CCC1C(=O)O)N
5 g
171560
97%

Chemical Identifiers

3685-25-4
143.186
DRNGLYHKYPNTEA-UHFFFAOYSA-N
171560
C1CC(CCC1C(=O)O)N
C7H13NO2
MFCD00209953
4-aminocyclohexanecarboxylic acid, trans-4-aminocyclohexanecarboxylic acid, cis-4-aminocyclohexanecarboxylic acid, cis-4-aminocyclohexane carboxylic acid, cis-4-amino-1-cyclohexanecarboxylic acid, trans-4-amino-cyclohexanecarboxylic acid, cis-4-aminocyclohexanecarboxylicacid, cyclohexanecarboxylic acid, 4-amino-, trans, trans-4-aminocyclohexane carboxylic acid, 1s,4s-4-aminocyclohexanecarboxylic acid
4-aminocyclohexane-1-carboxylic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only