missing translation for 'onlineSavingsMsg'
Learn More

trans-4-Amino-N-Boc-L-proline methyl ester hydrochloride, 97%

Catalog No. AAH5256603
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5256603 1 g
AAH5256606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5256603 Supplier Thermo Scientific Chemicals Supplier No. H5256603
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 334999-32-5 | C11H21ClN2O4 | 280.75 g/mol

trans-4-Amino-N-Boc-L-proline methyl ester hydrochloride acts as a pharmaceutical intermediate. It is used to prepare N-Boc-protected proline methyl ester of calix[4]arene, which is efficiently catalyzes the enantioselective aldol reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-4-Amino-N-Boc-L-proline methyl ester hydrochloride acts as a pharmaceutical intermediate. It is used to prepare N-Boc-protected proline methyl ester of calix[4]arene, which is efficiently catalyzes the enantioselective aldol reaction.

Notes
Store in a cool place. Moisture sensitive. Incompatible with oxidizing agents and acids.

Chemical Identifiers

CAS 334999-32-5
Molecular Formula C11H21ClN2O4
Molecular Weight (g/mol) 280.75
MDL Number MFCD03787929
InChI Key KFYCQKLSGMAVQH-WLYNEOFISA-N
Synonym 2s,4r-1-tert-butyl 2-methyl 4-aminopyrrolidine-1,2-dicarboxylate hydrochloride, n-boc-trans-4-amino-l-proline methyl ester hydrochloride, 2s,4r-1-boc-4-amino-l-proline methyl ester hcl, 2s,4r-4-amino-1-boc-pyrrolidine-2-carboxylic acid methyl ester-hcl, 1-tert-butyl 2-methyl 2s,4r-4-aminopyrrolidine-1,2-dicarboxylate hydrochloride, n-boc-trans-4-amino-l-proline methyl ester hydrochloride salt, methyl trans-3-amino-1-boc-5-pyrrolidinecarboxylate hcl, boc-trans-4-amino-l-proline methyl ester hydrochloride salt, 2s,4r-1-boc-4-amino-l-proline methyl ester hydrochloride, n-boc-trans-4-amino-l-proline methyl ester, hydrochloride salt
PubChem CID 17750455
SMILES [H+].[Cl-].COC(=O)[C@@H]1C[C@@H](N)CN1C(=O)OC(C)(C)C

Specifications

Quantity 1 g
IUPAC Name hydrogen 1-tert-butyl 2-methyl (2S,4R)-4-aminopyrrolidine-1,2-dicarboxylate chloride
Formula Weight 280.75
Percent Purity 97%
Chemical Name or Material trans-4-Amino-N-Boc-L-proline methyl ester hydrochloride
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only