accessibility menu, dialog, popup

UserName

CAS RN 334999-32-5

IUPAC Name: hydrogen 1-tert-butyl 2-methyl (2S,4R)-4-aminopyrrolidine-1,2-dicarboxylate chloride
Synonyms: hydrogen 1-tert-butyl 2-methyl (2S,4R)-4-aminopyrrolidine-1,2-dicarboxylate chloride
Molecular Weight (g/mol): 280.75
Molecular Formula: C11H21ClN2O4
InChi Key: KFYCQKLSGMAVQH-WLYNEOFISA-N
SMILES: [H+].[Cl-].COC(=O)[C@@H]1C[C@@H](N)CN1C(=O)OC(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results