missing translation for 'onlineSavingsMsg'
Learn More

trans-4-Acetoxycinnamic Acid 98.0+%, TCI America™

Catalog No. A28455G
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
A28455G 5 g
A284525G 25 g
2 options

Catalog No. A28455G

Supplier: TCI America A28455G

Only null left

Chemical Identifiers

CAS 27542-85-4
Molecular Formula C11H10O4
Molecular Weight (g/mol) 206.197
MDL Number MFCD00016847
InChI Key BYHBHNKBISXCEP-QPJJXVBHSA-N
Synonym trans-3-(4-Acetoxyphenyl)-2-propenoic Acid
PubChem CID 5373941
ChEBI CHEBI:86580
IUPAC Name (E)-3-(4-acetyloxyphenyl)prop-2-enoic acid
SMILES CC(=O)OC1=CC=C(C=C1)C=CC(=O)O

Specifications

Melting Point 210°C
Color White
Quantity 5 g
Formula Weight 206.20
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material trans-4-Acetoxycinnamic Acid