CAS RN 27542-85-4
CAS RN 27542-85-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
trans-4-Acetoxycinnamic Acid 98.0+%, TCI America™
CAS: 27542-85-4 Molecular Formula: C11H10O4 Molecular Weight (g/mol): 206.197 MDL Number: MFCD00016847 InChI Key: BYHBHNKBISXCEP-QPJJXVBHSA-N Synonym: trans-3-(4-Acetoxyphenyl)-2-propenoic Acid PubChem CID: 5373941 ChEBI: CHEBI:86580 IUPAC Name: (E)-3-(4-acetyloxyphenyl)prop-2-enoic acid SMILES: CC(=O)OC1=CC=C(C=C1)C=CC(=O)O