missing translation for 'onlineSavingsMsg'
Learn More

trans-4-(4-Chlorophenyl)cyclohexane-1-carboxylic acid, 98%

Catalog No. p-7032775
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3438414 25 g
AAH3438422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3438414 Supplier Thermo Scientific Chemicals Supplier No. H3438414

CAS: 49708-81-8 | C13H15ClO2 | 238.711 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 49708-81-8
Molecular Formula C13H15ClO2
Molecular Weight (g/mol) 238.711
MDL Number MFCD06797626
InChI Key NXXDIEYTMQYWJU-UHFFFAOYSA-N
Synonym 4-4-chlorophenyl cyclohexanecarboxylic acid, trans-4-4-chlorophenyl cyclohexanecarboxylic acid, 4-4-chlorophenyl cyclohexane-1-carboxylic acid, e-4-4-chlorophenyl cyclohexanecarboxylic acid, 4-4'-chlorophenyl cyclohexancarboxylic acid, cis-4-4-chlorophenyl cyclohexanecarboxylic acid, atovaquone related compound 1, cyclohexanecarboxylic acid, 4-4-chlorophenyl-, trans, 1r,4r-4-4-chlorophenyl cyclohexane-1-carboxylic acid, cyclohexanecarboxylic acid, 4-4-chlorophenyl
PubChem CID 2735792
IUPAC Name 4-(4-chlorophenyl)cyclohexane-1-carboxylic acid
SMILES C1CC(CCC1C2=CC=C(C=C2)Cl)C(=O)O

Specifications

Melting Point 249°C to 254°C
Quantity 25 g
Formula Weight 238.71
Percent Purity 98%
Chemical Name or Material trans-4-(4-Chlorophenyl)cyclohexane-1-carboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only