accessibility menu, dialog, popup

UserName

ClOHO

CAS RN 49708-81-8

IUPAC Name: 4-(4-chlorophenyl)cyclohexane-1-carboxylic acid
Synonyms: 4-(4-chlorophenyl)cyclohexane-1-carboxylic acid
Molecular Weight (g/mol): 238.71
Molecular Formula: C13H15ClO2
InChi Key: NXXDIEYTMQYWJU-UHFFFAOYSA-N
SMILES: OC(=O)C1CCC(CC1)C1=CC=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)