missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans-4'-(3,4-Difluorophenyl)bicyclohexyl-4-one 98.0+%, TCI America™
$145.40 - $426.87
Chemical Identifiers
| CAS | 147622-85-3 |
|---|---|
| Molecular Formula | C18H22F2O |
| Molecular Weight (g/mol) | 292.37 |
| InChI Key | IUJMYVYPSSBPCX-UHFFFAOYSA-N |
| PubChem CID | 11493045 |
| IUPAC Name | 4-[4-(3,4-difluorophenyl)cyclohexyl]cyclohexan-1-one |
| SMILES | C1CC(CCC1C2CCC(=O)CC2)C3=CC(=C(C=C3)F)F |
Chemical Identifiers
| 147622-85-3 | |
| 292.37 | |
| 11493045 | |
| C1CC(CCC1C2CCC(=O)CC2)C3=CC(=C(C=C3)F)F |
| C18H22F2O | |
| IUJMYVYPSSBPCX-UHFFFAOYSA-N | |
| 4-[4-(3,4-difluorophenyl)cyclohexyl]cyclohexan-1-one |
Specifications
| 147622-85-3 | |
| White | |
| 1 g | |
| C1CC(CCC1C2CCC(=O)CC2)C3=CC(=C(C=C3)F)F | |
| 292.37 | |
| 292.37 | |
| Crystalline Powder |
| 77°C | |
| C18H22F2O | |
| IUJMYVYPSSBPCX-UHFFFAOYSA-N | |
| 4-[4-(3,4-difluorophenyl)cyclohexyl]cyclohexan-1-one | |
| 11493045 | |
| ≥98.0% (GC) | |
| trans-4′-(3,4-Difluorophenyl)bicyclohexyl-4-one |
Safety and Handling
EINECSNumber : (4)-1815
TSCA : No