missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans-3-Pentenoic Acid 95.0+%, TCI America™
Supplier: TCI America P107225ML
Specifications
| trans-3-Pentenoic Acid | |
| Colorless | |
| C5H8O2 | |
| 25 mL | |
| UIUWNILCHFBLEQ-NSCUHMNNSA-N | |
| (3E)-pent-3-enoic acid | |
| 5282706 | |
| 100.12 | |
| Liquid |
| 1617-32-9 | |
| 93°C | |
| MFCD00063291 | |
| 3265 | |
| C\C=C\CC(O)=O | |
| 100.12 | |
| CHEBI:38370 | |
| ≥95.0% (GC,T) |
Chemical Identifiers
| 1617-32-9 | |
| 100.12 | |
| UIUWNILCHFBLEQ-NSCUHMNNSA-N | |
| CHEBI:38370 | |
| C\C=C\CC(O)=O |
| C5H8O2 | |
| MFCD00063291 | |
| 5282706 | |
| (3E)-pent-3-enoic acid |
Safety and Handling
EINECSNumber : (2)-0609
TSCA : No