accessibility menu, dialog, popup

UserName

CAS RN 1617-32-9

CH3HOO(E)

CAS RN 1617-32-9

IUPAC Name: (3E)-pent-3-enoic acid
Synonyms: trans-3-pentenoic acid trans-pent-3-ensaeure β-penteic acid pent-3t-enoic acid E-3-pentensaeure trans-β,gamma-penteneoic acid
Molecular Weight (g/mol): 100.12
Molecular Formula: C5H8O2
InChi Key: UIUWNILCHFBLEQ-NSCUHMNNSA-N
SMILES: C\C=C\CC(O)=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results