missing translation for 'onlineSavingsMsg'
Learn More

trans-3-Pentenoic Acid 95.0+%, TCI America™

Catalog No. P10725ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P10725ML 5 mL
P107225ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P10725ML Supplier TCI America Supplier No. P10725ML
Only null left

Chemical Identifiers

CAS 1617-32-9
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.12
MDL Number MFCD00063291
InChI Key UIUWNILCHFBLEQ-NSCUHMNNSA-N
PubChem CID 5282706
ChEBI CHEBI:38370
IUPAC Name (3E)-pent-3-enoic acid
SMILES C\C=C\CC(O)=O

Specifications

Color Colorless
Boiling Point 93°C
Quantity 5 mL
UN Number 3265
Formula Weight 100.12
Percent Purity ≥95.0% (GC,T)
Physical Form Liquid
Chemical Name or Material trans-3-Pentenoic Acid
EINECSNumber (2)-0609
TSCA No