missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans-3-Hexenedioic Acid 98.0+%, TCI America™
$146.31 - $413.92
Chemical Identifiers
| CAS | 4436-74-2 |
|---|---|
| Molecular Formula | C6H8O4 |
| Molecular Weight (g/mol) | 144.126 |
| MDL Number | MFCD00002785 |
| InChI Key | YHGNXQAFNHCBTK-OWOJBTEDSA-N |
| Synonym | trans-2-Butene-1,4-dicarboxylic Acid, Dihydromuconic Acid, trans-beta-Hydromuconic Acid |
| PubChem CID | 5351896 |
| ChEBI | CHEBI:86952 |
| IUPAC Name | (E)-hex-3-enedioic acid |
| SMILES | C(C=CCC(=O)O)C(=O)O |
Chemical Identifiers
| 4436-74-2 | |
| 144.126 | |
| YHGNXQAFNHCBTK-OWOJBTEDSA-N | |
| 5351896 | |
| (E)-hex-3-enedioic acid |
| C6H8O4 | |
| MFCD00002785 | |
| trans-2-Butene-1,4-dicarboxylic Acid, Dihydromuconic Acid, trans-beta-Hydromuconic Acid | |
| CHEBI:86952 | |
| C(C=CCC(=O)O)C(=O)O |
Specifications
| 4436-74-2 | |
| White | |
| MFCD00002785 | |
| trans-2-Butene-1,4-dicarboxylic Acid, Dihydromuconic Acid, trans-beta-Hydromuconic Acid | |
| C(C=CCC(=O)O)C(=O)O | |
| 144.126 | |
| CHEBI:86952 | |
| ≥98.0% (T) | |
| trans-3-Hexenedioic Acid |
| 198°C | |
| C6H8O4 | |
| 5 g | |
| YHGNXQAFNHCBTK-OWOJBTEDSA-N | |
| (E)-hex-3-enedioic acid | |
| 5351896 | |
| 144.13 | |
| Crystalline Powder |
Safety and Handling
TSCA : Yes