CAS RN 4436-74-2
CAS RN 4436-74-2
IUPAC Name:
(3E)-hex-3-enedioic acid
Synonyms:
3-hexenedioic acid
Molecular Weight (g/mol):
144.13
Molecular Formula:
C6H8O4
InChi Key:
YHGNXQAFNHCBTK-OWOJBTEDSA-N
SMILES:
OC(=O)C\C=C\CC(O)=O
1
–
1
of
1
results
1
trans-3-Hexenedioic Acid 98.0+%, TCI America™
CAS: 4436-74-2 Molecular Formula: C6H8O4 Molecular Weight (g/mol): 144.126 MDL Number: MFCD00002785 InChI Key: YHGNXQAFNHCBTK-OWOJBTEDSA-N Synonym: trans-2-Butene-1,4-dicarboxylic Acid, Dihydromuconic Acid, trans-beta-Hydromuconic Acid PubChem CID: 5351896 ChEBI: CHEBI:86952 IUPAC Name: (E)-hex-3-enedioic acid SMILES: C(C=CCC(=O)O)C(=O)O