missing translation for 'onlineSavingsMsg'
Learn More

trans-3-Hexene, 98%

Catalog No. p-4258993
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC149910010 1 g Glass bottle
AC149910051 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC149910010 Supplier Thermo Scientific Chemicals Supplier No. 149910010
Only null left

CAS: 13269-52-8 | C6H12 | 84.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13269-52-8
Molecular Formula C6H12
Molecular Weight (g/mol) 84.16
InChI Key ZQDPJFUHLCOCRG-AATRIKPKSA-N
Synonym trans-3-hexene, 3-hexene, e-3-hexene, 3-hexene, e, 3-hexene, 3e, unii-3co1lux1ar, e-hex-3-ene, 3co1lux1ar, alkenes, c6, cis-3-hexene
PubChem CID 638066
IUPAC Name (E)-hex-3-ene
SMILES CCC=CCC

Specifications

Melting Point -113.0°C
Density 0.6770g/mL
Boiling Point 67.0°C
Flash Point −12°C
Assay Percent Range 97.5% min
Packaging Glass bottle
Refractive Index 1.3935 to 1.3955
Linear Formula C2H5CH=CHC2H5
Quantity 1 g
Beilstein 01,II,192
Specific Gravity 0.677
Formula Weight 84.16
Percent Purity 98%
Chemical Name or Material trans-3-Hexene
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful if swallowed.
May be fatal if swallowed and enters airways.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 236-261-4

RUO – Research Use Only