accessibility menu, dialog, popup

UserName

CAS RN 13269-52-8

IUPAC Name: (3E)-hex-3-ene
Synonyms: trans-3-hexene
Molecular Weight (g/mol): 84.16
Molecular Formula: C6H12
InChi Key: ZQDPJFUHLCOCRG-AATRIKPKSA-N
SMILES: CC\C=C\CC
Boiling Point 
  • (2)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (4)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)