missing translation for 'onlineSavingsMsg'
Learn More

trans-3-Hexene, 98%

Catalog No. AAL1031106
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1031106 5 g
AAL1031103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1031106 Supplier Thermo Scientific Chemicals Supplier No. L1031106

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 13269-52-8 | C6H12 | 84.162 g/mol

Oligomers were prepared from the ozonolyses of cis- and trans-3-hexene and cis- and trans-2,5-dimethyl-3-hexene. Synthesis and characterization of aliphatic unsaturated polyesters is from trans-3-hexene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Oligomers were prepared from the ozonolyses of cis- and trans-3-hexene and cis- and trans-2,5-dimethyl-3-hexene. Synthesis and characterization of aliphatic unsaturated polyesters is from trans-3-hexene.

Solubility
Insoluble in water. Soluble in benzene, ligroin, ethanol, chloroform, ether.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 13269-52-8
Molecular Formula C6H12
Molecular Weight (g/mol) 84.162
MDL Number MFCD00009386
InChI Key ZQDPJFUHLCOCRG-AATRIKPKSA-N
Synonym trans-3-hexene, 3-hexene, e-3-hexene, 3-hexene, e, 3-hexene, 3e, unii-3co1lux1ar, e-hex-3-ene, 3co1lux1ar, alkenes, c6, cis-3-hexene
PubChem CID 638066
IUPAC Name (E)-hex-3-ene
SMILES CCC=CCC

Specifications

Melting Point -113°C
Density 0.677
Boiling Point 66°C to 67°C
Flash Point −25°C (−13°F)
Odor Mild
Refractive Index 1.394
Quantity 5 g
UN Number UN3295
Beilstein 1718859
Solubility Information Insoluble in water. Soluble in benzene,ligroin,ethanol,chloroform,ether.
Formula Weight 84.16
Percent Purity 98%
Chemical Name or Material trans-3-Hexene
Show More Show Less
Hazard Category H225-H302-H304
Hazard Statement GHS H Statement
H225-H304
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Precautionary Statement P210-P233-P240-P241-P242-P243-P264b-P270-P280-P301+P310-P303+P361+P353-P330-P331-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
EINECSNumber 236-261-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only