missing translation for 'onlineSavingsMsg'
Learn More

trans-3-chloroacrylic acid, 99%

Catalog No. AC108550010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC108550010 1 g Glass bottle
AC108550050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC108550010 Supplier Thermo Scientific Chemicals Supplier No. 108550010

CAS: 2345-61-1 | C3H3ClO2 | 106.51 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2345-61-1
Molecular Formula C3H3ClO2
Molecular Weight (g/mol) 106.51
MDL Number MFCD00064237
InChI Key MHMUCYJKZUZMNJ-OWOJBTEDSA-N
Synonym trans-3-chloroacrylic acid, e-3-chloroacrylic acid, 3-chloroacrylic acid, 3-chloroprop-2-enoic acid, acrylic acid, 3-chloro-, trans, acrylic acid, 3-chloro-, e, trans-3-chloropropenoic acid, trans-beta-chloroacrylic acid, 2-propenoic acid, 3-chloro-, e, ccris 3546
PubChem CID 638124
IUPAC Name (E)-3-chloroprop-2-enoic acid
SMILES C(=CCl)C(=O)O

Specifications

Melting Point 82°C to 85°C
Color Beige-Gray to Gray
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula ClCH=CHCO2H
Quantity 1 g
Beilstein 02, 400
Formula Weight 106.51
Percent Purity 99%
Physical Form Powder or Flakes
Chemical Name or Material trans-3-chloroacrylic acid
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 219-070-

RUO – Research Use Only