missing translation for 'onlineSavingsMsg'
Learn More

trans-3-Chloro-beta-nitrostyrene, 97%

Catalog No. p-7055465
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6405303 1 g
AAH6405306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6405303 Supplier Thermo Scientific Chemicals Supplier No. H6405303

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 37888-03-2 | C8H6ClNO2 | 183.591 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 37888-03-2
Molecular Formula C8H6ClNO2
Molecular Weight (g/mol) 183.591
MDL Number MFCD00175518
InChI Key GXQRAWTWDNHGBS-SNAWJCMRSA-N
Synonym 1-chloro-3-2-nitrovinyl benzene, e-1-chloro-3-2-nitrovinyl benzene, 1-3-chlorophenyl-2-nitroethene, trans-3-chloro-beta-nitrostyrene, beta-nitro-3-chlorostyrene, trans-1-chloro-3-2-nitro-vinyl-benzene, 1-chloro-3-e-2-nitrovinyl benzene, trans-1-3-chlorophenyl-2-nitroethene, cyto9f2, trans-3-chloro-, a-nitrostyrene
PubChem CID 5702314
IUPAC Name 1-chloro-3-[(E)-2-nitroethenyl]benzene
SMILES C1=CC(=CC(=C1)Cl)C=C[N+](=O)[O-]

Specifications

Melting Point 48°C to 52°C
Color Yellow
Quantity 1 g
Beilstein 2087804
Formula Weight 183.59
Percent Purity 97%
Physical Form Powder
Chemical Name or Material trans-3-Chloro-beta-nitrostyrene
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only