missing translation for 'onlineSavingsMsg'
Learn More

trans-3-(Boc-amino)cyclobutanecarboxylic acid, 97%, Thermo Scientific Chemicals

Catalog No. p-7041634
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6290203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6290203 Supplier Thermo Scientific Chemicals Supplier No. H6290203

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 939400-34-7 | Molecular Formula: C10H17NO4 | Molecular Weight: 215.249 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 939400-34-7
Molecular Formula C10H17NO4
Molecular Weight (g/mol) 215.249
MDL Number MFCD06657680
InChI Key KLCYDBAYYYVNFM-UHFFFAOYSA-N
Synonym cis-3-tert-butoxycarbonylamino cyclobutanecarboxylic acid, trans-3-tert-butoxycarbonylamino cyclobutanecarboxylic acid, 3-tert-butoxycarbonylamino cyclobutanecarboxylic acid, trans-3-tert-butoxycarbonyl amino cyclobutanecarboxylic acid, 3-boc-amino cyclobutanecarboxylic acid, 3-tert-butoxycarbonyl amino cyclobutanecarboxylic acid, cis-3-tert-butoxycarbonyl amino cyclobutanecarboxylic acid, cis-3-boc-amino cyclobutanecarboxylic acid, 3-tert-butoxy carbonyl amino cyclobutane-1-carboxylic acid, cis-3-boc-aminocyclobutanecarboxylic acid
PubChem CID 24720963
IUPAC Name 3-[(2-methylpropan-2-yl)oxycarbonylamino]cyclobutane-1-carboxylic acid
SMILES CC(C)(C)OC(=O)NC1CC(C1)C(=O)O

Specifications

Quantity 1 g
Formula Weight 215.24
Percent Purity 97%
Chemical Name or Material trans-3-(Boc-amino)cyclobutanecarboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only