accessibility menu, dialog, popup

UserName

CAS RN 939400-34-7

H3CH3CCH3OONHOHO

CAS RN 939400-34-7

IUPAC Name: 3-{[(tert-butoxy)carbonyl]amino}cyclobutane-1-carboxylic acid
Synonyms: 3-[(tert-butoxycarbonyl)amino]cyclobutane-1-carboxylic acid
Molecular Weight (g/mol): 215.25
Molecular Formula: C10H17NO4
InChi Key: KLCYDBAYYYVNFM-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NC1CC(C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results