missing translation for 'onlineSavingsMsg'
Learn More

trans-3,4,4-Trimethyl-2-pentene 98.0+%, TCI America™

Catalog No. t108101ml
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T108101ML 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T108101ML Supplier TCI America Supplier No. T10810.1ML
Only null left

Chemical Identifiers

CAS 39761-57-4
Molecular Formula C8H16
Molecular Weight (g/mol) 112.216
MDL Number MFCD00059236
InChI Key FZQMZRXKWHQJAG-VOTSOKGWSA-N
PubChem CID 5357254
IUPAC Name (E)-3,4,4-trimethylpent-2-ene
SMILES CC=C(C)C(C)(C)C

Specifications

Color Colorless
Quantity 0.1 mL
UN Number 3295
Formula Weight 112.22
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material trans-3,4,4-Trimethyl-2-pentene