CAS RN 39761-57-4
CAS RN 39761-57-4
IUPAC Name:
3,4,4-trimethylpent-2-ene
Synonyms:
2-pentene, 3,4,4-trimethyl-
Molecular Weight (g/mol):
112.22
Molecular Formula:
C8H16
InChi Key:
FZQMZRXKWHQJAG-UHFFFAOYSA-N
SMILES:
CC=C(C)C(C)(C)C
1
–
1
of
1
results
1
trans-3,4,4-Trimethyl-2-pentene 98.0+%, TCI America™
CAS: 39761-57-4 Molecular Formula: C8H16 Molecular Weight (g/mol): 112.216 MDL Number: MFCD00059236 InChI Key: FZQMZRXKWHQJAG-VOTSOKGWSA-N PubChem CID: 5357254 IUPAC Name: (E)-3,4,4-trimethylpent-2-ene SMILES: CC=C(C)C(C)(C)C